Learn More
Hach™ Test 'N Tube (TNT) Cuvette Tests
Description
Provides a self-contained package for mixing and measurement. Hach™ Test 'N Tube (TNT) Hach™ Test 'N Tube (TNT) Cuvette Tests include 16mm capped tubes and all necessary reagents in one convenient package for reliable results and easy handling.
Test 'N Tube products enable convenient testing with ready to use ampules and pre-measured reagents all packed into one convenient package. Available for determinations of a variety of parameters.
- Test 'N Tube: Capped 16mm vials provide a safe, self-contained package for mixing and measurement
- Powder Pillows: Premeasured, single dose packets of powdered reagent
- Dropper bottles: For easy addition of liquid reagents
- Precisely dosed reagents and test cuvettes for easy planning and inventory control
Specifications
Specifications
| Description | Colorimetric determination cuvette tests |
| For Use With (Application) | Nitrite Determination |
| For Use With (Equipment) | Hach DR900 |
| Includes | 16mm Tubes and all necessary reagents |
| No. of Tests | 50 |
| Quantity | 1 Kit |
| Range | 0.003 to 0.500mg/L NO2-N |
| Test Method | NitriVer 3 Diazotization |
| Type | Test 'N Tube |