Learn More
Mecmesin™ Test Stand for 59885-Series Force Gauge
Product Code. 11907979
Encompass
Change view
Click to view available options
Test Stand for 59885-Series Force Gauge
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11907979 Test Stand for 59885-Series Force Gauge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 11907979 Supplier Mecmesin™ Supplier No. 804001
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Accessory for 59885-Series force gauge.
  • Repeatable test results
  • Single-column motorized test stands
  • Crosshead speed: 1 to 1000mm/min
  • Single cycle test mode with full-speed crosshead return
  • Auto-travel reverse on alarm point or sample break
Specifications
Product Type Test Stand
Description Accessory for 59885-Series Force Gauge
For Use With (Application) Measuring force generated during compression