Learn More
Fisherbrand™ Test Tube Racks for 5 L Baths
Product Code. 17541906 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
16 x 16 to 19 mm Tubes
24 mm Tubes
30 x 10 to 13 mm Tubes
35 x 1.5 ml Tubes
44 x 0.5 mL Tubes
5 x 30 mm Tubes
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
17541906 5 x 30 mm Tubes Each
17521906 16 x 16 to 19 mm Tubes Each
17531906 24 mm Tubes Each
17511906 30 x 10 to 13 mm Tubes Each
17551906 35 x 1.5 ml Tubes Each
17561906 44 x 0.5 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17541906
Only null left
Add to basket
Add to basket
Test tube racks for use with Fisherbrand™ 5 L Digital Heated Circulating Baths.
  • Conveniently stores test tubes being used with Fisherbrand™ 5 L Digital Heated Circulating Baths
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Test Tubes
Holds 5 x 30 mm Tubes