missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone-16,16,17-d3 Pentanoate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Specifications
Specifications
| Chemical Name or Material | Testosterone-16,16,17-D3 17-Pentanoate |
| Molecular Formula | C24 D3 H33 O3 |
| Quantity | 0.01 g |
| Synonym | (17β)-17-[(1-Oxopentyl)oxy]androst-4-en-3-one-16,16,17-d3, Testosterone-d3 valerate, Testosterone-16,16,17-d3 17-valerate, Testosterone-16,16,17-d3 pentanoate, Testosterone-16,16,17-d3 17-pentanoate, Testosterone-16,16,17-D3 17-pentanoate |
| InChI Formula | InChI=1 S/C24H36O3/c1-4-5-6-22(26)27-21-10-9-19-18-8-7-16-15-17(25)11-13-23(16,2)20(18)12-14-24(19,21)3/h15,18-21 H,4-14H2,1-3H3/t18-,19-,20-,21-,23-,24-/m0/s1/i10D2,21 D |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]2(C)[C@@]1([2 H])OC(=O)CCCC |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-16,16,17-trideuterio-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-17-yl] pentanoate |
| Molecular Weight (g/mol) | 375.559 |
| Formula Weight | 375.285 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction