missing translation for 'onlineSavingsMsg'
Learn More

Testosterone, meets the requirements of Ph.Eur., Thermo Scientific Chemicals

Product Code. 10310504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10310504 25 g 25g
10042040 5 g 5g
10052040 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 10310504 Supplier Thermo Scientific Acros Supplier No. 164410250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Testosterone, CAS # 58-22-0, is a steroid sex hormone from the androstane family and it is important to regulate and maintain several biological functions.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Testosterone is a steroid sex hormone from the androstane family and is important for the maintenance of several biological functions

• This compound is an androgen that exerts its function by binding to and activating the androgen receptor

Applications

• Testosterone is irreversibly converted to dihydrotestosterone (DHT) in target tissues by the enzyme 5-∝ reductase

• Testosterone or DHT (its active metabolite) ligand-androgen receptor complexes act as a transcription factor, stimulating the expression of several responsive genes

• It antagonizes the androgen receptor to induce gene expression that causes the growth, development, and maintenance of several biological functions

• This compound is commonly used in vivo studies as an ester derivative due to its rapid metabolism by the liver

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-22-0
Molecular Formula C19H28O2
Molecular Weight (g/mol) 288.43
InChI Key MUMGGOZAMZWBJJ-DYKIIFRCSA-N
IUPAC Name (1S,3aS,3bR,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O

Specifications

Melting Point 153.0°C to 157.0°C
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9324
Specific Rotation +110.00
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in chloroform, alcohol, acetone, benzene,, dioxane and vegetable oils, slightly soluble in ether
Formula Weight 288.42
Chemical Name or Material Testosterone
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Testosterone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1A
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H360FD-May damage fertility. May damage the unborn child.
  • H351-Suspected of causing cancer.
  • H362-May cause harm to breast-fed children.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.