missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tet Repressor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA146294
Additional Details : Weight : 0.01000kg
Description
This antibody is not reactive to human or other mammalian proteins.
The Tet repressor protein (TetR) regulates transcription of a family of tetracycline resistance determinants in Gram-negative bacteria. The Tet system is the most widely used regulatory system for conditional gene expression in eukaryotic cells. TetR can be used for that purpose in some organisms without further modifications. In mammals and in a large variety of other organisms, however, eukaryotic transcriptional activator or repressor domains are fused to TetR to turn it into an efficient regulator.Specifications
Tet Repressor | |
Polyclonal | |
Unconjugated | |
pSC101 tetR; TetR | |
A synthetic peptide made to an internal portion of Escherichia coli TetR protein (between residues 25-75) | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1.0 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |