missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
208.00€ - 484.00€
Specifications
| Antigen | TET1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative), ICC/IF (Negative) |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18210176
|
Novus Biologicals
NBP1-78966SS |
0.025 mL |
208.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18258775
|
Novus Biologicals
NBP1-78966 |
0.1 mL |
484.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TET1 Polyclonal specifically detects TET1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC/IF (Negative).Specifications
| TET1 | |
| Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative), ICC/IF (Negative) | |
| Polyclonal | |
| Rabbit | |
| Mouse | |
| Q8NFU7 | |
| 80312 | |
| A synthetic peptide made to an internal portion of the human TET1 protein (between residues 1700 to 1800). [UniProt Q8NFU7] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| PBS, 30% Glycerol with 0.05% Sodium Azide | |
| bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1 | |
| TET1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title