Learn More
Invitrogen™ TET3 Recombinant Rabbit Monoclonal Antibody (HL3009)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556894
Product Code. 30275400
  • 500.00€ / 100µL
Description
TET3 Recombinant Monoclonal Antibody for Western Blot
TET3, a member of the ten-eleven-translocation (TET) family of genes, is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine and is most abundantly expressed in hematopoietic cells. Unlike the related TET2 protein, mutations in TET3 have not been observed in any myeloid malignancies. TET3 has been shown to be involved in the demethylation of zygotic DNA before the first mitosis and has been suggested to be involved in the epigenetic reprogramming of the zygotic paternal DNA following natural fertilization and may also contribute to somatic cell nuclear reprogramming during animal cloning.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TET3
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O43151
TET3
Recombinant fragment of human TET3.
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL3009
Unconjugated
TET3
B430006D22Rik; BC037432; D230004J03Rik; hCG_40738; KIAA0401; Methylcytosine dioxygenase TET3; MGC22014; probable methylcytosine dioxygenase TET3; putative methylcytosine dioxygenase; Tet methylcytosine deoxygenase 3 isoform; tet methylcytosine dioxygenase 3; tet oncogene family member 3; Tet3
Rabbit
Protein A
RUO
200424
-20°C, Avoid Freeze/Thaw Cycles