missing translation for 'onlineSavingsMsg'
Learn More

Tetra-Sodium Pyrophosphate Decahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10685692 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10685692 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10685692 Supplier Fisher Chemical Supplier No. S/6120/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tetra-Sodium Pyrophosphate Decahydrate, Extra Pure, SLR, Fisher Chemical, Quantity: 1kg, Packaging: Plastic powder jar, Boiling Point: 93.8 deg.C, CAS: 13472-36-1, White, Melting Point: 80 deg.C, Molecular Formula: H20Na4O17P2, Molecular Weight: 446.05, pH: 10

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 93.8°C
Melting Point 80°C
CAS 13472-36-1
Color White
pH 10
Physical Form Solid
Quantity 1 kg
Molecular Formula H20Na4O17P2
MDL Number 149200
Synonym sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
InChI Key VZWGHDYJGOMEKT-UHFFFAOYSA-J
SMILES O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
IUPAC Name tetrasodium;phosphonato phosphate;decahydrate
Molecular Weight (g/mol) 446.05
PubChem CID 3084150
Formula Weight 446.05g/mol
Grade Extra Pure
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • tetra-Sodium pyrophosphate decahydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium pyrophosphatedecahydrate, Sodiumpyrophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.