missing translation for 'onlineSavingsMsg'
Learn More

Tetraacetoxymethyl Bis(2-aminoethyl) Ether N,N,N’,N’-Tetraacetic Acid, TRC

Product Code. 30412979
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412979 50 mg -
30411730 10 mg -
30409572 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412979 Supplier Toronto Research Chemicals Supplier No. TRCT20000050MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887407-56-9
Molecular Formula C24H36N2O17
Molecular Weight (g/mol) 624.55
IUPAC Name 2-[carboxymethyl-[1,4-diacetyloxy-3-[1,4-diacetyloxy-3-[bis(carboxymethyl)amino]butan-2-yl]oxybutan-2-yl]amino]acetic acid
SMILES CC(=O)OCC(OC(COC(=O)C)C(COC(=O)C)N(CC(=O)O)CC(=O)O)C(COC(=O)C)N(CC(=O)O)CC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H36N2O17/c1-13(27)39-9-17(25(5-21(31)32)6-22(33)34)19(11-41-15(3)29)43-20(12-42-16(4)30)18(10-40-14(2)28)26(7-23(35)36)8-24(37)38/h17-20H,5-12H2,1-4H3,(H,31,32)(H,33,34)(H,35,36)(H,37,38)
Formula Weight 624.2014
Chemical Name or Material Tetraacetoxymethyl Bis(2-aminoethyl) Ether N,N,N',N'-Tetraacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.