missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromobisphenol S, TRC

Product Code. 30136754
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136754 100 g 100g
30138625 25 g 25g
17901005 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136754 Supplier Toronto Research Chemicals Supplier No. TRCT291110100G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39635-79-5
Molecular Formula C12 H6 Br4 O4 S
Molecular Weight (g/mol) 565.8546
Synonym 4,4'-Sulfonylbis[2,6-dibromophenol], 2,6-Dibromo-4-(3,5-dibromo-4-hydroxyphenyl)sulfonylphenol, 3,3',5,5'-Tetrabromobisphenol S, 3,5,3',5'-Tetrabromobisphenol S, 4,4'-Dihydroxy-3,5,3',5'-tetrabromodiphenyl sulfone, Bis(3,5-dibromo-4-hydroxyphenyl) sulfone, Bis(4-hydroxy-3,5-dibromophenyl) sulfone, EB 400S, FG 400S, FR 640, Flame Cut 160R, NFPP, Tetrabromobisphenol S
IUPAC Name 2,6-dibromo-4-(3,5-dibromo-4-hydroxyphenyl)sulfonylphenol
SMILES Oc1c(Br)cc(cc1Br)S(=O)(=O)c2cc(Br)c(O)c(Br)c2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C12H6Br4O4S/c13-7-1-5(2-8(14)11(7)17)21(19,20)6-3-9(15)12(18)10(16)4-6/h1-4,17-18H
Formula Weight 561.672
Chemical Name or Material Tetrabromobisphenol S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.