missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Tribromide, TRC

Product Code. 30416882
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416882 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416882 Supplier Toronto Research Chemicals Supplier No. TRCT29126010G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabutylammonium Tribromide
CAS 38932-80-8
Molecular Formula C16H36Br3N
Quantity 10 g
Synonym N,N,N-Tributyl-1-butanaminium Bromide, N,N,N,N-Tetrabutylammonium Tribromide
InChI Formula InChI=1S/C16H36N.Br3/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-3-2/h5-16H2,1-4H3;/q+1;-1
SMILES CCCC[N+](CCCC)(CCCC)CCCC.Br[Br-]Br
Molecular Weight (g/mol) 482.18
Formula Weight 479.0398
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.