missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylstannane, TRC

Product Code. 30135632
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135632 10 g 10g
30003735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135632 Supplier Toronto Research Chemicals Supplier No. TRCT29129010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-25-2
Molecular Formula C16 H36 Sn
Molecular Weight (g/mol) 347.17
Synonym Stannane, tetrabutyl-, Tetrabutylstannane, NSC 22330, NSC 28131, NSC 65524, Tetra-n-butylstannane, Tetra-n-butyltin, Tetrabutyltin, Tetrabutyltin(IV)
IUPAC Name tetrabutylstannane
SMILES CCCC[Sn](CCCC)(CCCC)CCCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/4C4H9.Sn/c4*1-3-4-2;/h4*1,3-4H2,2H3;
Formula Weight 348.1839
Chemical Name or Material Tetrabutyltin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.