missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracaine Hydrochloride 1.0 mg/ml in Acetonitrile (as free base), LoGiCal
Specifications
Specifications
| Chemical Name or Material | Tetracaine hydrochloride |
| CAS | 136-47-0 |
| Analyte or Component Names | Tetracaine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H24 N2 O2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Tetracaine Hydrochloride, Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-(butylamino)-, 2-(dimethylamino)ethyl ester, monohydrochloride, Benzoic acid, p-(butylamino)-, 2-(dimethylamino)ethyl ester, monohydrochloride, Ethanol, 2-dimethylamino-, p-butylaminobenzoate, HCl (4CI), 2-(Dimethylamino)ethyl p-(butylamino)benzoate hydrochloride, 2-Dimethylaminoethanol 4-n-butylaminobenzoate hydrochloride, Amethocaine hydrochloride, Amethocaine monohydrochloride, Anacel, Anethain, Anethaine, Butethanol, Curtacain, Decicain, Decicaine, Dessicain, Dianguang, Dicaine, Dikain, Intercaine, Leocaine, Menonasal, Niphanoid, Pantocain, Pantocaine, Pantocaine hydrochloride, Pantocaine monohydrochloride, Pontocaine hydrochloride, Tetocaine, Tetracaine chlorhydrate, Tetracaine hydrochloride, Tetracaine monohydrochloride, Tetracainium chloride, Tonexol, p-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester hydrochloride |
| InChI Formula | InChI=1S/C15H24N2O2.ClH/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3;/h6-9,16H,4-5,10-12H2,1-3H3;1H |
| Solution Type | Single Solution |
| SMILES | Cl.CCCCNc1ccc(cc1)C(=O)OCCN(C)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?