missing translation for 'onlineSavingsMsg'
Learn More

Tetraconazole, TRC

Product Code. 30137630
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137630 250 mg 250mg
30134774 100 mg 100mg
17934655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137630 Supplier Toronto Research Chemicals Supplier No. TRCT291475250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112281-77-3
Molecular Formula C13 H11 Cl2 F4 N3 O
Molecular Weight (g/mol) 372.15
Synonym 1H-1,2,4-Triazole, 1-[2-(2,4-dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]- (9CI, ACI), 1-[2-(2,4-Dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]-1H-1,2,4-triazole (ACI), 1H-1,2,4-Triazole, 1-[2-(2,4-dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]-, (±)- (ZCI), AG 4454, Domark, Emerald, Eminent, Greman, M 14360, Perissim, Salvatore ME, Tetraconazole
IUPAC Name 1-[2-(2,4-dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]-1,2,4-triazole
SMILES FC(F)C(F)(F)OCC(Cn1cncn1)c2ccc(Cl)cc2Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C13H11Cl2F4N3O/c14-9-1-2-10(11(15)3-9)8(4-22-7-20-6-21-22)5-23-13(18,19)12(16)17/h1-3,6-8,12H,4-5H2
Formula Weight 371.0215
Chemical Name or Material Tetraconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.