missing translation for 'onlineSavingsMsg'
Learn More

Tetracyclin Hydrochlorid, Honeywell Fluka™

Product Code. 16644494 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
16644494 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 16644494

Brand: Honeywell Fluka R1773100MG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetracyclin Hydrochlorid
CAS 64-75-5
CAS Max % ≤ 100%
Linear Formula C22H24N2O8 · HCl
Beilstein 3844873
Color Yellow
Melting Point 220°C to 225°C
Synonym (4S,4aS,5aS,6S,12aR)-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide hydrochloride
Molecular Weight (g/mol) 480.90 g/mol
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.