missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Tetracycline ReadyMade™ Solution

Product Code. 30044019
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044019 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044019 Supplier TOKU-E™ Supplier No. T14610x1ml
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tetracycline ReadyMade™ Solution is provided as a sterile-filtered solution of Tetracycline Hydrochloride (Tetracycline HCl) in water.

  • CAS Number: 64-75-5
  • Molecular Formula: C22H24N2O8 · HCl
  • Molecular Weight: 480.90 g/mol
  • Storage Conditions: -20°C, Protect from light
  • Form: Liquid
  • Source: Streptomyces sp.
  • Spectrum: Tetracycline is a broad-spectrum antibiotic, effective against both Gram-positive, Gram-negative bacteria, and mycoplasma. A few species of bacteria display intrinsic resistance to tetracycline, including Pseudomonas aeruginosa. The spectrum of Tetracycline activity encompasses various protozoa such as P. falciparum, Entamoeba histolytica, Giardia lamblia, Leishmania major, Trichomonas vaginalis, and Toxoplasma gondii
  • Mechanism of Action: Tetracycline inhibits bacterial growth in Gram-positive and Gram-negative bacteria by disrupting codon-anticodon interactions at the ribosome, thus blocking protein synthesis. Specifically, Tetracycline binds to a single site on the 30S ribosomal subunit and inhibit protein synthesis by blocking the attachment of charged aminoacyl-tRNA to the A site on the ribosome. They prevent introduction of new amino acids to the nascent peptide chain

Specifications

CAS 64-75-5
Certifications/Compliance Assay: Tetracycline dried substance: 95.0% to 102.0%
Chemical Name or Material Tetracycline Hydrochloride
Color Clear, colorless
Content And Storage -20°C, Protect from light
Description Tetracycline ReadyMade™ Solution 10 mg/mL in Water
Concentration 10.0 to 10.5 mg/mL
Form Clear, colorless solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in Water
For Use With (Application) Microbiology, Plant Biology, Eukaryotic Cell Culture and Cancer Applications
Product Line ReadyMade™ Solutions
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Source Streptomyces sp.
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.