missing translation for 'onlineSavingsMsg'
Learn More

Tetradecanal, TRC

Product Code. 30011530
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
Unit Size:
25g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011530 5 g 5g
30007816 500 mg 500mg
30008804 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011530 Supplier Toronto Research Chemicals Supplier No. TRCT2914405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-25-4
Molecular Formula C14 H28 O
Molecular Weight (g/mol) 212.37
Synonym Tetradecanal, 1-Tetradecanal, Aldehyde Peche, Myristinaldehyde, Myristylaldehyde, NSC 66435, Peach Aldehyde, Tetradecanaldehyde, Tetradecyl Aldehyde, n-Tetradecanal, n-Tetradecyl Aldehyde
IUPAC Name tetradecanal
SMILES CCCCCCCCCCCCCC=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H28O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h14H,2-13H2,1H3
Formula Weight 212.214
Chemical Name or Material Tetradecanal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.