missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetradecanamine, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Tetradecylamine |
| CAS | 2016-42-4 |
| Molecular Formula | C14 H31 N |
| Quantity | 25 g |
| Synonym | 1-Tetradecanamine (9CI','ACI), Tetradecylamine (6CI','8CI), 1-Aminotetradecane, 1-Tetradecylamine, Alamine 5D, Armeen 14, Armeen 14D, Farmin 40D, Monotetradecylamine, Myristylamine, n-Tetradecylamine, Nissan Amine MB, NSC 66437, Tetradecanamine |
| InChI Formula | InChI=1S/C14H31N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-15H2,1H3 |
| SMILES | CCCCCCCCCCCCCCN |
| IUPAC Name | tetradecan-1-amine |
| Molecular Weight (g/mol) | 213.4 |
| Formula Weight | 213.2457 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction