missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl ethylenediphosphonate, 98%

Product Code. 11462287
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11462287 5 g 5g
11452287 1 g 1g
11472287 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11462287

Brand: Thermo Scientific Alfa Aesar A14206.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 995-32-4 | C10H24O6P2 | 302.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 995-32-4
Molecular Formula C10H24O6P2
Molecular Weight (g/mol) 302.24
MDL Number MFCD00015133
InChI Key RSQYXXACEZCDFS-UHFFFAOYSA-N
Synonym tetraethyl ethylenediphosphonate, tetraethyl ethylenebisphosphonate, ethylenediphosphonic acid tetraethyl ester, tetraethyl ethane-1,2-diylbis phosphonate, 1,2-bis diethoxyphosphoryl ethane, 1,2-bis diethyl-phosphonato ethane, phosphonic acid, 1,2-ethanediylbis-, tetraethyl ester, diethyl 2-diethoxyphosphoryl ethylphosphonate, phosphonic acid, ethylenedi-, tetraethyl ester, acmc-209scy
PubChem CID 70446
IUPAC Name 1,2-bis(diethoxyphosphoryl)ethane
SMILES CCOP(=O)(CCP(=O)(OCC)OCC)OCC

Specifications

Density 1.146
Boiling Point 153°C to 154°C (0.9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.441
Quantity 5 g
Beilstein 1800096
Formula Weight 302.25
Percent Purity 98%
Chemical Name or Material Tetraethyl ethylenediphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.