missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium chloride monohydrate, 98+%

Product Code. 11387358
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387358 50 g 50g
11377358 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387358 Supplier Thermo Scientific Alfa Aesar Supplier No. 030818.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Phase transfer catalyst | CAS: 68696-18-4 | C8H22ClNO | 183.72 g/mol

Phase transfer catalystTetraethylammonium chloride monohydrate is used in the preparation of 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl chloride by reacting with 1,2-O-acetoxonium-3,4,6-tri-O-acetyl-alfaD-glucopyranose perchlorate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68696-18-4
Molecular Formula C8H22ClNO
Molecular Weight (g/mol) 183.72
MDL Number MFCD00149992
InChI Key CALLTGJPWMIDPC-UHFFFAOYSA-M
Synonym tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate
PubChem CID 16211205
IUPAC Name tetraethylazanium;chloride;hydrate
SMILES CC[N+](CC)(CC)CC.O.[Cl-]

Specifications

Melting Point ∼110°C
Density 1.0801 g/mL
Quantity 50 g
Beilstein 3563247
Merck Index 14,9200
Solubility Information Soluble in water.
Formula Weight 183.72 (165.71 Anhydrous)
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material Tetraethylammonium chloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.