missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Hydroxide, TRC

Product Code. 30449531
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449531 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449531 Supplier Toronto Research Chemicals Supplier No. TRCT303805500MG
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraethylammonium Hydroxide
CAS 77-98-5
Molecular Formula C8H21NO
Quantity 500 mg
Synonym Ethanaminium','N,N,N-triethyl-','hydroxide (1:1) (ACI), Ammonium','tetraethyl-','hydroxide (8CI), Ethanaminium','N,N,N-triethyl-','hydroxide (9CI), Tetraethylammonium hydroxide (6CI), EAH 20, TEAH 40W, N,N,N-Triethylethanaminium hydroxide
InChI Formula InChI=1S/C8H20N.H2O/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H2/q+1;/p-1
SMILES [OH-].CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium;hydroxide
Molecular Weight (g/mol) 147.26
Formula Weight 147.1623
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.