missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Hydroxide, TRC

Product Code. 30456156
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456156 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456156 Supplier Toronto Research Chemicals Supplier No. TRCT3038052.5G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetraethylammonium Hydroxide
CAS 77-98-5
Molecular Formula C8H21NO
Quantity 2.5 g
Synonym Ethanaminium','N,N,N-triethyl-','hydroxide (1:1) (ACI), Ammonium','tetraethyl-','hydroxide (8CI), Ethanaminium','N,N,N-triethyl-','hydroxide (9CI), Tetraethylammonium hydroxide (6CI), EAH 20, TEAH 40W, N,N,N-Triethylethanaminium hydroxide
InChI Formula InChI=1S/C8H20N.H2O/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H2/q+1;/p-1
SMILES [OH-].CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium;hydroxide
Molecular Weight (g/mol) 147.26
Formula Weight 147.1623
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.