missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroisophthalonitrile, 97%

Product Code. 11497214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497214 25 g 25g
11477214 1 g 1g
11487214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497214 Supplier Thermo Scientific Alfa Aesar Supplier No. B21599.14
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2377-81-3 | C8F4N2 | 200.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2377-81-3
Molecular Formula C8F4N2
Molecular Weight (g/mol) 200.10
MDL Number MFCD00013290
InChI Key WVHMPQKZPHOCRD-UHFFFAOYSA-N
Synonym tetrafluoroisophthalonitrile, 2,4,5,6-tetrafluoroisophthalonitrile, perfluoroisophthalonitrile, tetrafluorobenzene-1,3-dicarbonitrile, 1,3-benzenedicarbonitrile, 2,4,5,6-tetrafluoro, tetrafluoroisopthalonitrile, m-dicyanotetrafluorobenzene, acmc-209g5p, 1,3-dicyanoperfluorobenzene
PubChem CID 608182
SMILES FC1=C(F)C(C#N)=C(F)C(C#N)=C1F

Specifications

Melting Point 76°C to 78°C
Quantity 25 g
UN Number UN3439
Beilstein 6925899
IUPAC Name tetrafluorobenzene-1,3-dicarbonitrile
Formula Weight 200.1
Percent Purity 97%
Chemical Name or Material Tetrafluoroisophthalonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.