missing translation for 'onlineSavingsMsg'
Learn More

Tetrahexylammonium Iodide 99%, ACROS Organics™

Product Code. 10687401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
10g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10687401 10g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10687401 Supplier Thermo Scientific Acros Supplier No. 420420100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

CAS 2138-24-1
Melting Point 100°C to 105°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C24H52IN
Linear Formula (CH3(CH2)5)4NI
MDL Number MFCD00041981
Quantity 10 g
Synonym tetrahexylammonium iodide, tetra-n-hexylammonium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide, tetrahexylazanium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide 1:1, acmc-209fjk, tetrahexyl ammonium iodide, tetra-n-hexyl ammonium iodide, ammonium, tetrahexyl-, iodide, 1-hexanaminium,n,n-trihexyl-, iodide
InChI Key VRKHAMWCGMJAMI-UHFFFAOYSA-M
SMILES CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[I-]
IUPAC Name tetrahexylazanium;iodide
Molecular Weight (g/mol) 481.591
PubChem CID 75056
Formula Weight 481.58
Percent Purity ≥98.5%
Physical Form Fluffy Powder
Chemical Name or Material Tetrahexylammonium iodide, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.