missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrodicyclopentadiene, TRC

Product Code. 30457017
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457017 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457017 Supplier Toronto Research Chemicals Supplier No. TRCT795383500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrahydrodicyclopentadiene
CAS 6004-38-2
Molecular Formula C10H16
Quantity 500 mg
Synonym Octahydro-4,7-methano-1H-indene, 2,3-Trimethylenenorbornane, 2,3-(1,3-Propanediyl)bicyclo[2.2.1]heptane, Hexahydro-4,7-methanoindan, NSC 22464, Tetrahydrocyclopentadiene dimer, Tricyclo[5.2.1.02,6]decane, Trimethylenenorbornane
InChI Formula InChI=1S/C10H16/c1-2-9-7-4-5-8(6-7)10(9)3-1/h7-10H,1-6H2
SMILES C1(CCC2)C2C3CC1CC3
Molecular Weight (g/mol) 136.23
Formula Weight 136.1252
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.