missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl Acrylate, TRC

Product Code. 30419430
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30419430

Brand: Toronto Research Chemicals TRCT204660250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrahydrofurfuryl Acrylate
CAS 2399-48-6
Molecular Formula C8 H12 O3
Quantity 250 mg
InChI Formula InChI=1S/C8H12O3/c1-2-8(9)11-6-7-4-3-5-10-7/h2,7H,1,3-6H2
SMILES C=CC(=O)OCC1CCCO1
IUPAC Name oxolan-2-ylmethyl prop-2-enoate
Molecular Weight (g/mol) 156.179
Formula Weight 156.079
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.