missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofurfuryl Acrylate, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
Specifications
Specifications
| Chemical Name or Material | Tetrahydrofurfuryl Acrylate |
| CAS | 2399-48-6 |
| Molecular Formula | C8 H12 O3 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C8H12O3/c1-2-8(9)11-6-7-4-3-5-10-7/h2,7H,1,3-6H2 |
| SMILES | C=CC(=O)OCC1CCCO1 |
| IUPAC Name | oxolan-2-ylmethyl prop-2-enoate |
| Molecular Weight (g/mol) | 156.179 |
| Formula Weight | 156.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction