missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydroxydiboron, 95%

Product Code. 16504214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16504214 5 g 5g
16514214 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16504214 Supplier Thermo Scientific Acros Supplier No. 461280050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13675-18-8 | B2H4O4 | 89.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13675-18-8
Molecular Formula B2H4O4
Molecular Weight (g/mol) 89.65
MDL Number MFCD05663888
InChI Key SKOWZLGOFVSKLB-UHFFFAOYSA-N
IUPAC Name (dihydroxyboranyl)boronic acid
SMILES OB(O)B(O)O

Specifications

Assay Percent Range 95%
Linear Formula (HO)2BB(OH)2
Quantity 5 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol
Formula Weight 89.65
Chemical Name or Material Tetrahydroxydiboron
compliance-icons
Product Identifier
  • Tetrahydroxydiboron, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydroxydiboron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.