Learn More
Tetrahydroxydiboron, 95%
Product Code. 16514214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16514214 25 g 25g
16504214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16514214 Supplier Thermo Scientific Acros Supplier No. 461280250
Request Bulk or Custom Format
CAS: 13675-18-8 | B2H4O4 | 89.65 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13675-18-8
Molecular Formula B2H4O4
Molecular Weight (g/mol) 89.65
MDL Number MFCD05663888
InChI Key SKOWZLGOFVSKLB-UHFFFAOYSA-N
IUPAC Name (dihydroxyboranyl)boronic acid
SMILES OB(O)B(O)O
Specifications
Assay Percent Range 95%
Linear Formula (HO)2BB(OH)2
Quantity 25 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol
Formula Weight 89.65
Chemical Name or Material Tetrahydroxydiboron
compliance-icons
Product Identifier
  • Tetrahydroxydiboron, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydroxydiboron