missing translation for 'onlineSavingsMsg'
Learn More

Tetraisopropyl Methylenediphosphonate, TRC

Product Code. 30428008
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428008 25 g -
30416421 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428008 Supplier Toronto Research Chemicals Supplier No. TRCT29627025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660-95-3
Molecular Formula C13 H30 O6 P2
Molecular Weight (g/mol) 344.32
Synonym P,P'-Methylenebisphosphonic acid, Phosphonic acid, methylenebis-, tetrakis(1-methylethyl) ester (9CI), Phosphonic acid, methylenedi-, tetraisopropyl ester (7CI,8CI), Methylenediphosphonic acid tetraisopropyl ester, NSC 226577, Tetraisopropyl methylenebisphosphonate, Tetraisopropyl methylenediphosphonate, Tetrakis(O-isopropyl) methylenediphosphonate, [(Diisopropoxyphosphinyl)methyl]diisopropoxyphosphine oxide
IUPAC Name 2-[di(propan-2-yloxy)phosphorylmethyl-propan-2-yloxyphosphoryl]oxypropane
SMILES CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H30O6P2/c1-10(2)16-20(14,17-11(3)4)9-21(15,18-12(5)6)19-13(7)8/h10-13H,9H2,1-8H3
Formula Weight 344.1518
Chemical Name or Material Tetraisopropyl methylenediphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.