missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(decyl)ammonium Bromide, TRC

Product Code. 30409453
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409453 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409453 Supplier Toronto Research Chemicals Supplier No. TRCT29643050G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetradecylammonium Bromide
CAS 14937-42-9
Molecular Formula C40 H84 N . Br
Quantity 50 g
Synonym Tetrakis(decyl)ammonium Bromide, 1-Decanaminium','N,N,N-tris(decyl)-','bromide (1:1) (ACI), 1-Decanaminium','N,N,N-tris(decyl)-','bromide (9CI), Ammonium','tetrakis(decyl)-','bromide (8CI), Tetrakis(decyl)ammonium bromide (7CI), Terakis(decyl)ammonium bromide, Tetra-n-decylammonium bromide, Tetradecylammonium bromide, N,N,N-Tris(decyl)-1-decanaminium bromide
InChI Formula InChI=1S/C40H84N.BrH/c1-5-9-13-17-21-25-29-33-37-41(38-34-30-26-22-18-14-10-6-2,39-35-31-27-23-19-15-11-7-3)40-36-32-28-24-20-16-12-8-4;/h5-40H2,1-4H3;1H/q+1;/p-1
SMILES [Br-].CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC
IUPAC Name tetrakis-decylazanium;bromide
Molecular Weight (g/mol) 659.006
Formula Weight 657.579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.