missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)diboron, Thermo Scientific™

Product Code. 17302551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17302551 5 g 5g
17312551 25 g 25g
17322551 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17302551 Supplier Thermo Scientific Acros Supplier No. 468540050
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1630-79-1
Molecular Formula C8H24B2N4
Molecular Weight (g/mol) 197.93
MDL Number MFCD00048012
InChI Key KMCDRSZVZMXKRL-UHFFFAOYSA-N
IUPAC Name dimethyl[1,2,2-tris(dimethylamino)diboran-1-yl]amine
SMILES CN(C)B(B(N(C)C)N(C)C)N(C)C

Specifications

Melting Point -33°C
Density 0.926
Boiling Point 55°C to 57°C (2.5 mmHg)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Refractive Index 1.4680 to 1.4760 (20°C, 589 nm)
Quantity 5 g
Formula Weight 197.93
Physical Form Liquid
Chemical Name or Material Tetrakis(dimethylamino)diboron
compliance-icons compliance-icons
Product Identifier
  • Tetrakis(dimethylamino)diboron
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Tetrakis(dimethylamino)diboron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.