Learn More
Tetrakis(dimethylamino)titanium, 99.99%, (trace metal basis)
Product Code. 10642223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10642223 25 g Glass bottle 25g
10068701 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10642223 Supplier Thermo Scientific Acros Supplier No. 353560250
 Restricted Item
In Stock
Add to basket
CAS: 3275-24-9 | C8H24N4Ti | 224.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3275-24-9
Molecular Formula C8H24N4Ti
Molecular Weight (g/mol) 224.18
MDL Number MFCD00014861
InChI Key MNWRORMXBIWXCI-UHFFFAOYSA-N
Synonym tetrakis dimethylamino titanium, tetrakis dimethylamino titanium iv, tdmat, tetrakis dimethylamido titanium iv, tetrakis dimethylamido titanium, methanamine, n-methyl-, titanium 4+ salt, titanium dimethylamide, methanamine, n-methyl-, titanium 4+ salt 4:1, titanium tetrakis dimethylamide, acmc-209hva
PubChem CID 123185
SMILES [Ti+4].C[N-]C.C[N-]C.C[N-]C.C[N-]C
Specifications
Color Yellow to Orange
Density 0.9470g/mL
Flash Point -30°C
Infrared Spectrum Authentic
Assay Percent Range 99.99%
Packaging Glass bottle
Linear Formula [(CH3)2N]4Ti
Quantity 25 g
Beilstein 04, IV, 128
Specific Gravity 0.947
IUPAC Name dimethylazanide;titanium(4+)
Formula Weight 224.21
Percent Purity 99.99%
Grade Trace Metal Basis
Physical Form Liquid
Chemical Name or Material Tetrakis(dimethylamino)titanium
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrakis(dimethylamino)titanium
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.