missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium iodide, 99%, ACROS Organics™

Product Code. 10608634 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10608634 500 g Glass bottle 500g
10022870 25 g Glass bottle 25g
10226800 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10608634 Supplier Thermo Scientific Acros Supplier No. 216685000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-58-1
Molecular Formula C4H12IN
Molecular Weight (g/mol) 201.05
MDL Number MFCD00011629
InChI Key RXMRGBVLCSYIBO-UHFFFAOYSA-M
Synonym tetramethylammonium iodide, artilacer, yodurtan, banikol, tmai, methanaminium, n,n,n-trimethyl-, iodide, ammonium, tetramethyl-, iodide, methanaminium, n,n,n-trimethyl-, iodide 1:1, tetramethylammoniumiodide, tetramethylammoniumjodid
PubChem CID 6381
IUPAC Name tetramethylazanium iodide
SMILES [I-].C[N+](C)(C)C

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)4NI
Quantity 500 g
Beilstein 04, 51
Merck Index 15, 9372
Formula Weight 201.05
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetramethylammonium iodide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.