missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium iodide, 99%

Product Code. 11435183
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435183 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11435183 Supplier Thermo Scientific Alfa Aesar Supplier No. A12811.36
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-58-1 | C4H12IN | 201.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-58-1
Melting Point >300°C
Odor Odorless
Molecular Formula C4H12IN
Linear Formula (CH3)4NI
MDL Number MFCD00011629
Quantity 500 g
UN Number UN2811
Beilstein 3620030
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9225
Synonym tetramethylammonium iodide, artilacer, yodurtan, banikol, tmai, methanaminium, n,n,n-trimethyl-, iodide, ammonium, tetramethyl-, iodide, methanaminium, n,n,n-trimethyl-, iodide 1:1, tetramethylammoniumiodide, tetramethylammoniumjodid
Solubility Information Slightly soluble aq. soln.; Insoluble in chloroform, ether
InChI Key RXMRGBVLCSYIBO-UHFFFAOYSA-M
SMILES [I-].C[N+](C)(C)C
IUPAC Name tetramethylazanium;iodide
Molecular Weight (g/mol) 201.05
PubChem CID 6381
Formula Weight 201.05
Percent Purity 99%
Chemical Name or Material Tetramethylammonium iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.