missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylol Acetylenediurea, TRC

Product Code. 30132848
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132848 10 g 10g
30132407 5 g 5g
30008477 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132848 Supplier Toronto Research Chemicals Supplier No. TRCT88970810G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5395-50-6
Molecular Formula C8 H14 N4 O6
Molecular Weight (g/mol) 262.22
Synonym Glycoluril, 1,3,4,6-tetrakis(hydroxymethyl)- (6CI,7CI,8CI), Tetrahydro-1,3,4,6-tetrakis(hydroxymethyl)imidazo[4,5-d]imidazole-2,5(1H,3H)-dione, 1,3,4,6-Tetrakis(hydroxymethyl)glycoluril, 2,4,6,8-Tetrakis(hydroxymethyl)glycoluril, 2,5-Dioxo-1,3,4,6-tetrakis(hydroxymethyl)hexahydroimidazo[4,5-d]imidazole, Cymel 1172, Fixapret 140, N,N',N' ',N' ' '-Tetrakis(hydroxymethyl)glycoluril, NSC 3319, NSC 67452, Protectol TD, Tetrahydro-1,3,4,6-tetrakis(hydroxymethyl)imidazo[4,5-d]imidazol-2,5(1H,3H)-dione, Tetrakis(hydroxymethyl)acetylenediurea, Tetrakis(hydroxymethyl)acetyleneurea, Tetrakis(hydroxymethyl)glycoluril, Tetramethylol acetylene carbamide, Tetramethylolacetylenediurea, Tetramethylolglycoluril
IUPAC Name 1,3,4,6-tetrakis(hydroxymethyl)-3a,6a-dihydroimidazo[4,5-d]imidazole-2,5-dione
SMILES OCN1C2C(N(CO)C(=O)N2CO)N(CO)C1=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H14N4O6/c13-1-9-5-6(11(3-15)7(9)17)12(4-16)8(18)10(5)2-14/h5-6,13-16H,1-4H2
Formula Weight 262.0913
Chemical Name or Material Tetrakis(hydroxymethyl)glycoluril
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.