missing translation for 'onlineSavingsMsg'
Learn More

Tetraxolin, TRC

Product Code. 30134987
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134987 250 mg 250mg
30138107 500 mg 500mg
17902505 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134987 Supplier Toronto Research Chemicals Supplier No. TRCN756885250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100343-43-9
Molecular Formula C10 H8 O6
Molecular Weight (g/mol) 224.167
Synonym 2,3-Dihydro-2,2,3,3-tetrahydroxy-1,4-naphthalenedione;2,2,3,3-Tetrahydroxynaphthalene-1,4-dione;Oksolin, Oxolin, Oxoline;
IUPAC Name 2,2,3,3-tetrahydroxytetralin-1,4-dione
SMILES OC1(O)C(=O)c2ccccc2C(=O)C1(O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H8O6/c11-7-5-3-1-2-4-6(5)8(12)10(15,16)9(7,13)14/h1-4,13-16H
Formula Weight 224.0321
Chemical Name or Material Tetraxolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.