missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgJ Antibody (OTI3B3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71016AF700
This item is not returnable.
View return policy
Description
IgJ Monoclonal specifically detects IgJ in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| IgJ | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| JCHAIN | |
| Full length recombinant protein of human IgJ (NP_653247) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI3B3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| IGCJJCH, immunoglobulin J chain, immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mupolypeptides | |
| Mouse | |
| Affinity Purified | |
| Immunology | |
| 3512.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction