Learn More
Texwipe™ Sterile Polystyrene Swabs
Product Code. 17278879 Shop All Texwipe Products
Change view
Click to view available options
Width (English) Head:
0.22 in.
Width (Metric) Head:
5.8 mm
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Head Width (Metric) Head Unit Size
17278879 0.22 in. 5.8 mm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17278879 Supplier Texwipe™ Supplier No. STX763
  • ONLINE EXCLUSIVE PRICE
    1088.52€
    803.39€ / Pack of 500
    Save 285.13€
    26% Off
Estimated Shipment: 02-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Texwipe’s Sterile Spun Polyester Swabs are constructed using 100% USP-grade polyester, securely bonded to the wooden or polystyrene handle by a water-based adhesive.
  • Environmental monitoring
  • Cleaning with compatible solutions and solvents
  • Picking up fine powders
  • Surface sampling
  • Collection and transport of bacteriological specimens
  • Specifications
    Length (English) 6.1 in.
    Product Type Swab
    Length (Metric) 155 mm
    Material Medical-grade spun polyester
    Width (English) Head 0.22 in.
    Width (Metric) Head 5.8 mm
    Handle Type Polystyrene
    Adhesive Type Water-based
    Handle Material Polystyrene
    Tip Style Large
    Tip Material Foam