missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Antibody (CL14112) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26853B
This item is not returnable.
View return policy
Description
TFEB Monoclonal antibody specifically detects TFEB in Human samples. It is validated for Immunohistochemistry
Specifications
| TFEB | |
| Monoclonal | |
| Biotin | |
| ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
| This antibody was developed using a synthetic peptide derived from P19484, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Autophagy | |
| 7942 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry | |
| CL14112 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction