missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF2 Antibody (SR1241), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22379-100ul
This item is not returnable.
View return policy
Description
TFF2 Monoclonal antibody specifically detects TFF2 in Human samples. It is validated for Western Blot
Specifications
| TFF2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7032 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR1241 | |
| Western Blot 1:500-1:2000 | |
| SML1trefoil factor 2, SML1, human spasmolytic polypeptide (SP)10spasmolytic protein 1, SP, spasmolysin, Spasmolytic polypeptide, trefoil factor 2 | |
| A synthesized peptide derived from human TFF2 (Uniprot #: Q03403) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction