missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF beta 1/2/3 Antibody (1D11), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1835U
This item is not returnable.
View return policy
Description
TGF beta 1/2/3 Monoclonal antibody specifically detects TGF beta 1/2/3 in Broad Species Reactivity samples. It is validated for Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Specifications
| TGF beta 1/2/3 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7040 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Binding Inhibition Assay, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| 1D11 | |
| Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor, beta 1 | |
| Bovine bone-derived TGF-beta 1 and TGF-beta 2 | |
| 0.1 mL | |
| Primary | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction