missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1 Antibody (MM0446-6G14) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11766-0.025MG
This item is not returnable.
View return policy
Description
TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.
Specifications
| TGF-beta 1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 1:10-1:1000, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready | |
| P01137 | |
| TGFB1 | |
| Human TGF beta 1 | |
| 0.025 mg | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology | |
| 7040 | |
| Human | |
| Purified |
| Flow Cytometry, Blocking Assay, CyTOF | |
| MM0446-6G14 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction