Learn More
TGFBI / BIGH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10188-1-AP-150UL
 View more versions of this product
Product Code. 16836403
  • 410.00€ / 150µL
Description
This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy.
Specifications
Specifications
TGFBI / BIGH3
Polyclonal
Unconjugated
TGFBI
Beta ig h3, BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, Kerato epithelin, LCD1, RGD CAP, TGFBI
Rabbit
Antigen Affinity Chromatography
RUO
116487, 21810, 7045
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P82198, Q15582
TGFBI
TGFBI / BIGH3 Fusion Protein Ag0241
150 μL
Primary
Human, Mouse, Rat