missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12912-3-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody recognizes the endogenous 90 kDa TGM1 proteins.
The protein encoded by this gene is a membrane protein that catalyzes the addition of an alkyl group from an akylamine to a glutamine residue of a protein, forming an alkylglutamine in the protein. This protein alkylation leads to crosslinking of proteins and catenation of polyamines to proteins. This gene contains either one or two copies of a 22 nt repeat unit in its 3′ UTR. Mutations in this gene have been associated with autosomal recessive lamellar ichthyosis (LI) and nonbullous congenital ichthyosiform erythroderma (NCIE).Spezifikation
| TGM1 | |
| Polyclonal | |
| Unconjugated | |
| TGM1 | |
| Epidermal TGase, ICR2, KTG, LI, LI1, TG(K), TGASE, TGase 1, TGase K, TGK, TGM1, Transglutaminase 1, Transglutaminase K | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21816, 60335, 7051 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P22735, P23606, Q9JLF6 | |
| Tgm1 | |
| TGM1 Fusion Protein Ag3905 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur