missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTCO1 Antibody (3.7C4) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43547
This item is not returnable.
View return policy
Description
MTCO1 Monoclonal antibody specifically detects MTCO1 in Mouse samples. It is validated for ELISA
Specifications
| MTCO1 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA | |
| COI, COX1, MTCO1, MT-COI, MT-COI mitochondrially encoded cytochrome c oxidase I | |
| This antibody was raised by immunising OVA-primed mice with MTCO1-OVA. (Uniprot# P00397) | |
| 0.2 mg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4512 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 3.7C4 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction