missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGM7 Monoclonal Antibody (602708), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524272
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant human TGM2, 3, 4, or recombinant mouse TGM2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
TGM7 Monoclonal antibody specifically detects TGM7 in Human samples. It is validated for Western Blot.Specifications
TGM7 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96PF1 | |
TGM7 | |
Sf 21-derived recombinant human Transglutaminase 7/TGM7 Asp2-Pro710 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot | |
602708 | |
Unconjugated | |
TGM7 | |
LOC102552199; Protein-glutamine gamma-glutamyltransferase Z; protein-glutamine gamma-glutamyltransferase Z-like; TG(Z); TGase Z; TGase-7; TGM7; TGMZ; TGMZEC 2.3.2.13; TGz; transglutaminase 7; transglutaminase Z; Transglutaminase-7 | |
Rat | |
Protein A/G | |
RUO | |
116179 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |