Learn More
Tocris Bioscience™ TH 10785
Description
TH 10785 is an 8-oxo guanine DNA glycosylase 1 (OGG1) activator. It binds in the active site of OGG1 and increases its recruitment to oxidative DNA damage and its repair.
Specifications
Specifications
| Chemical Name or Material | N-Cyclohexyl-2-cyclopropyl-4-quinazolinamine |
| CAS | 1002801-51-5 |
| Quantity | 10 mg |
| Target | Glycosylase Activators |
| Molecular Formula | C17H21N3 |
| Purity | 0.98 |