missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Thalidomide-Cyanine 5
GREENER_CHOICE

Product Code. 18787246
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18787246 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18787246 Supplier Tocris Bioscience™ Supplier No. 7288/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity cereblon fluorescent probe

Thalidomide-Cyanine 5 is a fluorescent probe for cereblon E3 ligases. Binds with high affinity to DDB1-CRBN. Suitable for use in TR-FRET.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-((1 E,3 E)-5-((E)-1-(6-((4-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamido)butyl)amino)-6-oxohexyl)-3,3-dimethyl-5-sulfoindolin-2-ylidene)penta-1,3-dien-1-yl)-1-ethyl-3,3-dimethyl-3 H-indol-1-ium-5-sulfonate
Quantity 1 mg
Target Fluorescent Receptor Probes
Molecular Formula C52H60N6O13S2
Purity 0.9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.